Learn More
R&D Systems™ Mouse CLEC-1 (NP_780735) VersaClone cDNA
Description
- Also known as: 5930406N14Rik
- CLEC-1 is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily.
- Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cellcell signaling, glycoprotein turnover, and roles in inflammation and immune response.
- CLEC-1 may play a role in regulating dendritic cell function.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec1a |
| Accession Number | NP_780735 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 823 bp |
| Gene ID (Entrez) | 51267 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |