missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Ceruloplasmin ELISA Kit (Colorimetric)

Product Code. 18612581 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18612581

Brand: Novus Biologicals™ NBP282160

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.13 to 200 ng/mL
Assay Sensitivity 1.88 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Specificity This kit recognizes Mouse Ceruloplasmin in samples. No Significant cross-reactivity or interference between Mouse Ceruloplasmin and analogues was observed.
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 1356
Gene Symbol CP
Quantity 1 Kit
Research Discipline Cardiovascular Biology, Neurodegeneration, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ceruloplasmin, ceruloplasmin (ferroxidase), CP-2, EC 1.16.3.1, Ferroxidase
Target Mouse
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.