missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL11/Eotaxin Quantikine ELISA Kit
Click to view available options
Quantity:
2 x 96-well Plates
Unit Size:
1 set
Description
The Mouse CCL11/Eotaxin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL11/Eotaxin in cell culture supernates (50 ul), serum (13 ul), edta plasma (13 ul), citrate plasma (13 ul).
Specifications
Specifications
| Assay Range | 15.60 - 1000pg/mL |
| Assay Sensitivity | 3pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum |
| Specificity | Natural and Recombinant Mouse Eotaxin. This assay cross-reacts 0.062% with Human Eotaxin and recognizes Rat Eotaxin. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20292 |
| Quantity | 2 x 96-well Plates |
| Sample Volume | Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Citrate Plasma (13uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction