Learn More
Abcam Mouse brain tissue lysate - mitochondrial extract (contains SDS/DTT), 50 ug
Product Code. 32149238 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32149238 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32149238 Supplier Abcam Supplier No. ab11034550ug
 Restricted Item
Estimated Shipment: 09-12-2026
Mouse brain cell/tissue lysate.
  • Species: Mouse
  • Tissue: brain
  • Form: Liquid
Specifications
For Use With (Application) SDS-PAGE, Western Blot