missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse BCMA/TNFRSF17 DuoSet ELISA
Description
The Mouse BCMA/TNFRSF17 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse BCMA/TNFRSF17.
Specifications
Specifications
| Quantity | 15 x 96-well Plates |
| Conjugate | Biotin |
| Assay Range | 78.13 to 5000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 21935 |
| Synonym | B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?