missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse BAFF / TNFSF13B / CD257 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
457.25€ - 3357.52€
Specifications
Accession Number | NP_296371.1 |
---|---|
Molecular Weight (g/mol) | 47.7 kDa |
Source | Mouse BAFF, Mouse IgG2a Fc Tag, low endotoxin (BAF-M5257) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1). |
Immunogen | Ala 127 - Leu 309 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15836007
|
ACROBiosystems
BAF-M5257-1MG |
1 mg |
3357.52€
1mg |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
15826007
|
ACROBiosystems
BAF-M5257-100UG |
100 μg |
457.25€
100µg |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
Description
B-cell activating factor (BAFF) is also known as tumor necrosis factor ligand superfamily member 13B , TNFSF13B, BAFF, B Lymphocyte Stimulator (BLyS) , cluster of differentiation 257 (CD257), DTL, TNF- and APOL-related leukocyte expressed ligand (TALL-1), THANK, TNFSF20, ZTNF4, and is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells. It is expressed as transmembrane protein on various cell types including monocytes, dendritic cells and bone marrow stromal cells. BAFF is the natural ligand of three unusual tumor necrosis factor receptors named BAFF-R, TACI, and BCMA, all of which have differing binding affinities for it. These receptors are expressed mainly on mature B lymphocytes (TACI is also found on a subset of T-cells and BCMA on plasma cells). TACI binds worst since its affinity is higher for a protein similar to BAFF, called a proliferation-inducing ligand (APRIL). BCMA displays an intermediate binding phenotype and will work with either BAFF or APRIL to varying degrees. Signaling through BAFF-R and BCMA stimulates B lymphocytes to undergo proliferation and to counter apoptosis. All these ligands act as heterotrimers (i.e. three of the same molecule) interacting with heterotrimeric receptors, although BAFF has been known to be active as either a hetero- or homotrimer. BAFF acts as a potent B cell activator and has been shown to play an important role in the proliferation and differentiation of B cells. This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_296371.1 | |
Mouse BAFF, Mouse IgG2a Fc Tag, low endotoxin (BAF-M5257) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1). | |
-20°C | |
BAFF | |
Mouse | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
47.7 kDa | |
Ala 127 - Leu 309 | |
TNFSF13B,BAFF,BLYS,CD257,DTL,TALL1,THANK,TNFSF20,ZTNF4,TALL-1 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |