missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Artemin DuoSet ELISA

Product Code. 30000598
Change view
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000598 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000598 Supplier R&D Systems™ Supplier No. DY108505
  • ONLINE EXCLUSIVE PRICE
    501.00€
    473.55€ / Pack of 5
    Save 27.45€
    5% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Mouse Artemin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Artemin.
TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 0pg/mL
Detectable Analytes Artemin
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
For Use With (Application) ELISA
Gene ID (Entrez) 11876
Host Species Mouse
Measuring Range 31.2-2,000pg/mL
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.