missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgA, Clone: A93-2, Secondary Antibody, BD
Brand: BD Biosciences 553912
Additional Details : Weight : 0.01000kg
Description
The A93-2 antibody reacts specifically with rat IgA. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: A93-2
Isotype: IgG1, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgA
ELISA
Specifications
Mouse anti-Rat IgA | |
A93-2 | |
Biotin | |
0.5 mg | |
Immunology | |
Rat |
Monoclonal | |
0.5mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG1 κ |