missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas Ligand/TNFSF6 Mouse anti-Human, Clone: 100419, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
Brand: R&D Systems MAB126-100
This item is not returnable.
View return policy
Description
Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Human samples. It is validated for Flow Cytometry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| Fas Ligand/TNFSF6 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 1-5 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P48023 | |
| FASLG | |
| Chinese hamster ovary cell line CHO-derived recombinant human Fas Ligand/TNFSF6 Pro134-Leu281 Accession # P48023 | |
| 100 μg | |
| Primary | |
| Detects human Fas Ligand/TNFSF6 in ELISAs. | |
| Human | |
| Supernatant |
| Neutralization, ELISA | |
| 100419 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 356 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction