missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Chicken Ig Mix Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
440.00€ - 876.00€
Specifications
Antigen | Chicken Ig Mix |
---|---|
Clone | CIG10-183.12 |
Concentration | 0.6 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17890486
|
Invitrogen
SA5-10351 |
100μg |
876.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11594990
|
Invitrogen
SA110645 |
500 μg |
440.00€
1mg |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
17800446
|
Invitrogen
SA5-10350 |
1 mg |
876.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
SA1-10645 detects chicken Immunoglobulin. Does not react with bovine, canine, donkey, equine, feline, rabbit, rat, sheep, porcine, guinea pig, goat, or human samples. SA1-10645 has been successfully used in ELISA applications. The SA1-10645 immunogen is purified chicken immunoglobulin.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Chicken Ig Mix | |
0.6 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
IgG1 | |
Protein G |
CIG10-183.12 | |
ELISA | |
Unconjugated | |
Mouse | |
Chicken | |
Purified chicken immunoglobulin. | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles |