missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Canine Ig Mix Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
470.00€ - 876.00€
Specifications
Antigen | Canine Ig Mix |
---|---|
Clone | DIG11-124.1 |
Concentration | 0.9 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17830236
|
Invitrogen
SA5-10369 |
100μg |
876.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11574980
|
Invitrogen
SA110633 |
500 μg |
470.00€
1mg |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
17880306
|
Invitrogen
SA5-10368 |
1 mg |
876.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
SA1-10633 detects canine Immunoglobulin. This antibody shows no cross-reactivity with bovine, equine, feline, rabbit, ovine, donkey, porcine, guinea pig, goat, or human immunoglobulin proteins. SA1-10633 has been successfully used in ELISA applications. The SA1-10633 immunogen is purified canine immunoglobulin.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Canine Ig Mix | |
0.9 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
IgG1 | |
Protein A |
DIG11-124.1 | |
ELISA | |
Unconjugated | |
Mouse | |
Canine | |
Purified canine immunoglobulin. | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles |