Learn More
Mouse anti-Alix, Clone: 3A9, Alexa Fluor 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65678AF488
Encompass
Product Code. 15733586
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Alix Monoclonal specifically detects Alix in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.