Learn More
R&D Systems™ Mouse Activin RIA/ALK-2 (NP_001103674) VersaClone cDNA
Specifications
Specifications
| Gene | Activin RIA/ALK-2/Activin Receptor Type 1 |
| Accession Number | NP_001103674 |
| Gene ID (Entrez) | 90 |
| Gene Alias | activin A receptor, type I, ActivinRIA, ACTRI, ACVR1, ACVR1A, ACVRLK2, ALK2, ALK-2, FOP, SKR1, TSRI |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |