Learn More
MP Biomedicals™ Mounting Medium, Immuno-Fluore
Product Code. 11429025 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429025 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11429025 Supplier MP Biomedicals™ Supplier No. 08622701
designed for immunofluorescent preparations
TRUSTED_SUSTAINABILITY
Specifications
Product Type Immuno-Fluore mounting medium
Quantity 25 mL
For Use With (Application) Immunofluorescence (IF)
Form Liquid
Content And Storage 15° to 30°C