Learn More
asecos™ Mounting Kit for Floor Anchoring of Depots
Product Code. 17806596 Shop All Asecos Products
Click to view available options
Weight (English):
2.78 lb.
Unit Size:
Each
Product Code. 17806596
Brand: asecos™ 31859
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Mounting Kit is ideal for anchoring depots to the floor.
  • Compatible with all depots.
Specifications
Material Steel
Type Mounting Kit
Weight (English) 2.78 lb.
Weight (Metric) 1.26 kg