missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Mount Support

Product Code. 11825600
Click to view available options
Type:
Mount S with 76 mm dia. Support
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11825600

Brand: Zeiss™ 4354020000000

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Designed for use with Zeiss Microscopes

Compatible Microscopes:

  • Axio Zoom.V16
  • Stemi 2000
  • Stemi 508
  • Stemi 508 Demo
  • Stemi 508 doc
  • Stemi 508 trino
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12

Specifications

Type Mount S with 76 mm dia. Support
Diameter (Metric) Support: 76mm
For Use With (Equipment) Zeiss Microscopes
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.