Learn More
Heinz Herenz Mortar Pressed Glass
Product Code. 30158401
Change view
Click to view available options
Diameter (Metric) Outer:
80 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
30158401 80 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30158401 Supplier Heinz Herenz Supplier No. 1070580
  • ONLINE EXCLUSIVE PRICE
    645.75€
    522.46€ / Pack of 12
    Save 123.29€
    19% Off
Estimated Shipment: 20-11-2026
Mortar with 80 mm outer diameter. Pressed glass.
  • Mortar, pressed glass
  • 80 mm outer diameter
  • Porcelain
Specifications
Autoclavable Autoclavable
Diameter (Metric) Outer 80 mm
Product Type Mortar and Pestle
Material Glass
Finish Unglazed
Capacity (Metric) 80 mm