Learn More
Mono-tert-butyl Succinate, TRC
Specifications
Specifications
| CAS | 15026-17-2 |
| Molecular Formula | C8 H14 O4 |
| Quantity | 1 g |
| Synonym | tert-Butyl Succinate, 4-tert-Butoxy-4-oxobutanoic Acid, tert-Butyl Succinic Acid Ester, Mono-tert-Butyl Succinic Acid Ester, mono(1,1-Dimethylethyl)butanedioic Acid Ester, (1,1-Dimethylethyl)butanedioic Acid Ester |
| InChI Formula | InChI=1S/C8H14O4/c1-8(2,3)12-7(11)5-4-6(9)10/h4-5H2,1-3H3,(H,9,10) |
| SMILES | CC(C)(C)OC(=O)CCC(=O)O |
| IUPAC Name | 4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 174.194 |
| Formula Weight | 174.089 |
| Chemical Name or Material | Mono-tert-butyl Succinate |