Learn More
mono-n-Propyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Monopropyl Phthalate-d4 |
| CAS | 1795020-91-5 |
| Molecular Formula | C11 D4 H8 O4 |
| Quantity | 0.05 g |
| Synonym | 2,3,4,5-Tetradeuterio-6-propoxycarbonylbenzoic acid, mono-Propyl Phthalate-3,4,5,6-d4, Monopropyl Phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid monopropyl ester, Phthalic acid-d4 monopropyl ester, Propyl hydrogen phthalate-d4, Monopropyl Phthalate-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C11H12O4/c1-2-7-15-11(14)9-6-4-3-5-8(9)10(12)13/h3-6 H,2,7H2,1H3,(H,12,13)/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCC)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-propoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 212.235 |
| Formula Weight | 212.099 g/mol |
| Show More |