missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-n-Butyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Monobutyl Phthalate-D4 |
| CAS | 478954-81-3 |
| Molecular Formula | C12 2H4 H10 O4 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C12H14O4/c1-8(2)7-16-12(15)10-6-4-3-5-9(10)11(13)14/h3-6,8 H,7H2,1-2H3,(H,13,14)/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCC(C)C)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(2-methylpropoxycarbonyl)benzoic acid |
| Molecular Weight (g/mol) | 226.26 |
| Formula Weight | 226.1143 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction