Learn More
Cole Parmer Mono-Mold Standard Stir Bar with Spinning Ring
Shop All Cole-Parmer Products
Click to view available options
Length (English):
3 in.
Diameter (English):
1/2 in.
Unit Size:
Each
Description
Mono-Mold Standard Stir Bar is an all-purpose stir bar.
- Available with or without spinning rings. Rings reduce contact area between stir bar and vessel, reducing friction and noise.
- Super-strong alnico V magnets remain coupled to drive magnets - resisting spin-out at high speeds.
- Chemically inert
- Low-friction
- Leakproof PTFE coating provides smooth spinning and maximum energy transfer
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (English) | 1/2 in. |
| Includes | Spinning ring |
| Length (English) | 3 in. |
| Coating | PTFE |
| Diameter (Metric) | 12.7 mm |
| Length (Metric) | 76.2 mm |
| Material | PTFE |
| Type | Stir Bar |