missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-Cyclohexyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Monocyclohexyl Phthalate-d4 |
| CAS | 1398066-18-6 |
| Molecular Formula | C14 D4 H12 O4 |
| Quantity | 0.05 g |
| Synonym | mono-Cyclohexyl Phthalate-3,4,5,6-d4, Monocyclohexyl Phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid monocyclohexyl ester, Phthalic acid-d4 monocyclohexyl ester, Cyclohexyl hydrogen phthalate-d4, Monocyclohexyl Phthalate-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C14H16O4/c15-13(16)11-8-4-5-9-12(11)14(17)18-10-6-2-1-3-7-10/h4-5,8-10 H,1-3,6-7H2,(H,15,16)/i4D,5 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC2CCCCC2)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2-cyclohexyloxycarbonyl-3,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 252.299 |
| Formula Weight | 252.13 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction