missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-Benzyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Monobenzyl Phthalate-d4 |
| CAS | 478954-83-5 |
| Molecular Formula | C15 2H4 H8 O4 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C15H12O4/c16-14(17)12-8-4-5-9-13(12)15(18)19-10-11-6-2-1-3-7-11/h1-9 H,10H2,(H,16,17)/i4D,5 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCc2ccccc2)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-phenylmethoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 260.28 |
| Formula Weight | 260.0987 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction