Learn More
Mono(4-carboxybutyl) Phthalate, TRC
Specifications
Specifications
| Chemical Name or Material | Mono(4-carboxybutyl) Phthalate |
| CAS | 92569-48-7 |
| Molecular Formula | C13 H13 O6 |
| Quantity | 25 mg |
| Synonym | 1,2-Benzenedicarboxylic Acid 1-(4-Carboxybutyl) Ester, MCBP, 1,2-Benzenedicarboxylic Acid Mono(4-carboxybutyl) Ester |
| InChI Formula | InChI=1S/C13H14O6/c14-11(15)7-3-4-8-19-13(18)10-6-2-1-5-9(10)12(16)17/h1-2,5-6H,3-4,7-8H2,(H,14,15)(H,16,17)/p-1 |
| SMILES | OC(=O)CCCCOC(=O)c1ccccc1C(=O)[O-] |
| IUPAC Name | 2-(4-carboxybutoxycarbonyl)benzoate |
| Molecular Weight (g/mol) | 265.239 |
| Formula Weight | 265.072 |