missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Molybdenum Reagent Sets, For Colorimetric Water Testing
Description
These high quality reagents are designed to follow an adaptation of the mercaptoacetic acid method for colorimetric determination of molybdenum. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the yellow color that is produced.
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Specifications
| Analytes | Mercaptoacetic acid |
| For Use With (Application) | Molybdenum determination |
| For Use With (Equipment) | Compatible Hanna benchtop and portable photometers |
| Includes | 100 individual packets |
| No. of Tests | 100 |
| Quantity | 1 Set |
| Range | 0 to 2mg/L molybdenum |
| Test Method | Cholorimetric determination |
| Type | Water Testing Kit |
Safety and Handling
Product Identifier
- X100 TEST MOLYBDEN FOR 38935
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362-Take off contaminated clothing.
- P370+P378-In case of fire: Use powder to extinguish.
- P390-Absorb spillage to prevent material damage.
- P404-Store in a closed container.
- MIXTURE LIST-Contain: EDTA, succinic acid,potassium persulphate, calcium thioglycolate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?