Learn More
Agencinox™ Molybdene Stainless Steel Beakers without Spout
Product Code. p-8008856
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. 11751893
Brand: Agencinox™ 2050
Only null left
Add to basket
Add to basket
Use these beakers for a variety of general laboratory applications. Agencinox™ Molybdenum Stainless Steel Beakers ensure the safe transfer of liquids.
  • Stainless steel Molybdenum construction
  • High chemical resistance
  • Available in a variety of sizes
Specifications
Diameter (Metric) Outer 80 mm
Capacity (Metric) 500 mL
Material Stainless Steel (Molybdenum)
Height (Metric) 110 mm
Type Beaker