Learn More
Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Shop All MBP Molecular BioProducts Products
Click to view available options
Color:
Assorted
Green
Orange
Sterility:
Sterile
Capacity (Metric):
0.6 mL
Unit Size:
Pack of 10000
Description
- With flat cap
- No frosting or graduations
- Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
Specifications
Specifications
| Cap Style | Flat |
| Certifications/Compliance | RNase, DNase, RNA, DNA, ATP, pyrogen-free |
| Capacity (Metric) | 0.6 mL |
| Color | Orange |
| Material | Polypropylene |
| Includes | Flat cap |
| Packaging | Bag |
| Sterility | Sterile |
| Graduated | No |
| Closure Type | Flat Cap |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur