missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Undergoes reverse osmosis
- Meets USP and EPTesting specifications
- DNase and RNase-free
- Suitable for cell-culture and molecular biology applications
- Long-storage when handled properly under defined conditions
- pH: 5.0 to 7.0
- Appearance: Clear Solution
- Endotoxins: test < 0.05 EU/mL
- DNase detection: Not Detected
- RNase detection: Not Detected
Specifications
Specifications
| Chemical Name or Material | Water |
| Molecular Weight (g/mol) | Unknown |
| Percent Purity | Unknown |
| Grade | Cell Culture/Molecular Biology |
| Packaging | Bottle |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?