missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MolBoolean Mouse/Rabbit Assay Kit

Product Code. 31852700 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1.6 mL
Unit Size:
1.60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31852700 1.6 mL 1.60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31852700 Supplier Novus Biologicals™ Supplier No. MolB000011.6ml
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For analyzing protein proximity within cells and tissues using advanced immunofluorescent detection.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1.6 mL
Target MolBoolean Mouse/Rabbit
For Use With (Application) Proximity Ligation Assay
Product Type Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.