missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modifying Enzymes

Product Code. 31722960 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
DNase I
T4 RNA Ligase
T4 RNA Ligase 2
Vaccinia Capping Enzyme
Vaccinia mRNA Cap 2'-O-Methyltransferase
Quantity:
1000 U
1500 U
20,000 U
4000 U
5000 U
Concentration:
10 U/μL
30 U/μL
50 U/μL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Concentration unitSize
31722960 T4 RNA Ligase 1500 U 30 U/μL Each
31722880 Vaccinia mRNA Cap 2'-O-Methyltransferase 20,000 U 50 U/μL Each
31722726 Vaccinia Capping Enzyme 4000 U 10 U/μL Each
31722724 T4 RNA Ligase 2 1000 U 10 U/μL Each
31722812 DNase I 5000 U 50 U/μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31722960 Supplier Thermo Scientific™ Supplier No. EL002AF
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
TRUSTED_SUSTAINABILITY

Specifications

Concentration 30 U/μL
For Use With (Application) RNA Ligation
Content And Storage 30 U/μL T4 RNA Ligase, 50 μL

Store at -20°C.
Compatible Buffer T4 RNA Ligase Reaction Buffer
Sensitive to Heat Inactivation Yes
Quantity 1500 U
Product Type T4 RNA Ligase
Research Category RNA Research
Inactivation Temperature 70°C
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.