Learn More
Corning™ Modified Impeller Assembly
Shop All Corning Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Magnetically-Driven 15L Proculture Spinner Flask
Magnetically-Driven 3L Proculture Spinner Flask
Magnetically-Driven 8L Proculture Spinner Flask
missing translation for 'unitSize'
Each
Description
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
Specifications
Specifications
| Description | Impeller shaft with dual bearing |
| Material | Stainless Steel |
| For Use With (Equipment) | Magnetically-Driven 3L Proculture Spinner Flask |
| Sterility | Non-sterile |