Learn More
Memmert™ MobileALERT For Up to 4 Alarm Notifications
Product Code. 17110025 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Product Code. 17110025
Brand: Memmert™ ICO50C4
Only null left
Add to basket
Add to basket
MobileALERT for 2 alarm notifications, temperature and humidity
  • Accessory
  • Notification by SMS, temperature and CO2 alarm standard
  • Additionally humidity alarm when equipped with option K7
  • O2 alarm when equipped with option T6
Specifications
Description Temperature and humidity, up to 4 alarms
For Use With (Equipment) ICO50