Learn More
Köttermann Mobile Single Bench
This Mobile Single Bench fulfills a variety of tasks in the laboratory.
Brand:  Köttermann 505-00147
Additional Details : Weight : 38.62340kg
 View more versions of this product
Product Code. 17342209
  • 1120.74€ / Each
Estimated Shipment: 23-08-2024
Description
  • Available for sitting and standing work height
  • Features castors for mobility
Specifications
Specifications
750 x 900 x 750 mm
Melamine TopResist™ Worktop