Learn More
Köttermann Mobile Cabinet with Two Doors
Click to view available options
No. of Drawers:
1
2
None
Dimensions (D x W x H):
516 x 900 x 590 mm
516 x 900 x 740 mm
516 x 1200 x 590 mm
516 x 1200 x 740 mm
Unit Size:
Each
Description
- Versatile storage space
- Mobile with lockable wheels
- Features two doors to protect stored items
- Many sizes and configurations available
Specifications
Specifications
| Color | White |
| Cabinet Type | Mobile |
| Dimensions (D x W x H) | 516 x 900 x 590 mm |
| Door Type | Hinged |
| Finish | Epoxy Coated |
| Frame Material | Steel |
| Hinge Side | Left and Right |
| No. of Doors | 2 |
| No. of Drawers | None |
| No. of Shelves | None |
| Show More |