Learn More
asecos™ Mobile Box Sorbents Refill Kit
Shop All Asecos Products
Click to view available options
Type:
Chemical Spill Control Absorbent Storage Cart Refill Kit
Oil Spill Control Absorbent Storage Cart Refill Kit
Universal All Purpose Absorbent Storage Cart Refill Kit
Unit Size:
Each
Description
- content: 6 cushions, 3 socks, 20 mats
Specifications
Specifications
| Material | Heavy-duty Cloth |
| Type | Oil Spill Control Absorbent Storage Cart Refill Kit |
| Weight (English) | 6.61 lb. |
| Weight (Metric) | 3 kg |