Learn More
Tocris Bioscience™ ML 339
Potent and selective hCXCR6 antagonist
Brand: Tocris Bioscience™ 5943/10
This item is not returnable.
View return policy
Description
ML 339 is a potent and selective hCXCR6 antagonist (IC50 = 140 nM); 100-fold less active at the murine CXCR6 receptor (IC50 = 18 μM). Exhibits selectivity over CXCR5, CXCR4, CCR6 and APJ receptors (IC50 >79 μM).Specifications
| N-[(1 R,3 S,5 S)-9-(2-((2-Chlorophenyl)amino)-2-exoethyl)-9-azabicyclo[3.3.1 ]nonan-3-yl)-3,4,5-trimethoxybenzamide | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
| 2080300-49-6 | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| White |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.