Learn More
Tocris Bioscience™ ML 339
GREENER_CHOICE
Potent and selective hCXCR6 antagonist
Brand:  Tocris Bioscience™ 5943/10
Encompass
Product Code. 15732158
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
ML 339 is a potent and selective hCXCR6 antagonist (IC50 = 140 nM); 100-fold less active at the murine CXCR6 receptor (IC50 = 18 μM). Exhibits selectivity over CXCR5, CXCR4, CCR6 and APJ receptors (IC50 >79 μM).
Specifications
Specifications
N-[(1 R,3 S,5 S)-9-(2-((2-Chlorophenyl)amino)-2-exoethyl)-9-azabicyclo[3.3.1 ]nonan-3-yl)-3,4,5-trimethoxybenzamide
10 mg
Solid
Bioactive Small Molecule
2080300-49-6
Soluble to 100mM in DMSO and to 20mM in ethanol
White