missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ mL 334

Product Code. 18094422
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094422 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18094422 Supplier Tocris Bioscience™ Supplier No. 5625/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of Keap1- Nrf2 interaction

mL 334 is an Nrf2 activator; inhibits Nrf2/Keap1 interaction (Kd = 1 μM). Promotes Nrf2 nuclear translocation and induces antioxidant response element (ARE) activity in vitro. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S,2R)-2-[[(1S)-1-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-3,4-dihydro-2(1H)-isoquinolinyl]carbonyl]cyclohexanecarboxylic acid
CAS 1432500-66-7
Quantity 5 mg
Target Nrf2 Activators
Molecular Formula C26H26N2O5
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.