Learn More
Kinematica™ Mixing Attachment for Microtron™ MB 550 Laboratory Mixer
Product Code. 11960359 Shop All Kinematica Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11960359 125 mL Each
10637693 250 mL Each
11950049 500 mL Each
11960049 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11960359 Supplier Kinematica™ Supplier No. 36050005
Estimated Shipment: 03-12-2026
For the mixing, dispersing, homogenizing, emulsifying and size reduction of cuttable materials.
  • For use with the MB 550 Mixers
  • The two stainless steel ball bearings are sealed twice each
  • Noiseless and sturdy coupling with dampers
  • Blades, sealing and dampers can easily be exchanged
  • Four high-grade blades designed for optimal mixing
  • Generally suited for intensive and fast processing
Order Info
dimCapacityMetric
Specifications
Product Type Mixing Head Attachment
Capacity (Metric) 125 mL
Material Borosilicate Glass
Description Consists of gasket and blade head
For Use With (Equipment) MB 550 Laboratory Mixer
Packaging Quantity 1 Ea.