Learn More
Cytiva™ Whatman™ ME27/41 ST Mixed Cellulose Ester (MCE) Filtration Membranes, with Grid
Click to view available options
Diameter (Metric):
50 mm
Unit Size:
Pack of 100
Description
- Product Type: Filtration Membrane
- Grade: ME27/41 ST
- Material: Mixed Cellulose Ester (MCE; cellulose acetate and nitrocellulose)
- Diameter: 50 mm
- Pore Size: 0.8μm
- Grid Pattern: Black on Green (3.1 mm)
- For Use With (Application): Large Sample Volumes and Low Microorganism Counts
- Quantity: 100 Pk.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Green |
| Disposable | Reusable |
| Material | MCE |
| Pore Size | 0.8 μm |
| Sterility | Sterile |
| Flow Rate | 2.8/100/12.5 s/mL/cm2 |
| For Use With (Application) | Large Sample Volumes and Low Microorganism Counts |
| Gas Flow Rate | 11.6/100 s/mL |
| Grade | ME27/41 ST |
| Show More |
Safety and Handling
Signal Word
- Warning
- Flammable solid Category 1
- Flammable solid Category 2
- H228-Flammable solid.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : nitrocellulose