Learn More
Tocris Bioscience™ MitoBrilliant 646
Product Code. 30040316
Change view
Click to view available options
Quantity:
1 Pk. (5 x 50 μg)
50 μg (Trial size)
Unit Size:
250µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040316 50 μg (Trial size) 50µg
30564781 1 Pk. (5 x 50 μg) 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30040316 Supplier Tocris Bioscience™ Supplier No. 7700/50U
Universal red fluorescent mitochondrial stain for both live and fixed cells
MitoBrilliant probes have been developed to overcome some common limitations encountered with standard mitochondrial trackers, offering clearer answers to scientific questions. They are next-generation fluorescent stains for the localization and tracking of mitochondria in both live and fixed cells. The range harnesses Janelia Fluor® dye technology, conferring some of the properties that make these widely used dyes, into mitochondrial stains.
MitoBrilliant™ 646 is a next-generation universal stain for mitochondria in both live and fixed cells. It is retained in mitochondria following fixation with exceptionally clear staining and is retained in the mitochondria of live cells following loss of the mitochondrial membrane potential. Additionally, MitoBrilliant™ 646 can be used to stain pre-fixed cells/tissue, making this dye suitable for IHC/ICC.
Two dyes that accumulate in the mitochondria of live cells in a mitochondrial membrane potential (Δψm) dependent manner are also available: MitoBrilliant™ Live 646 (red emission) and MitoBrilliant™ Live 549 (yellow/orange emission).
Please refer to the protocol for guidelines on product use, and download the MitoBrilliant Product Guide to view data for each of the dyes in different applications.
MitoBrilliant is a trademark of Bio-Techne Corp.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C
Quantity 50 μg (Trial size)
Target Mitochondria Dyes and Probes