Learn More
Tocris Bioscience™ Miro1 Reducer
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Miro1 Reducer is a promotes proteasomal degradation of Miro1 (mitochondrial Rho GTPase 1). Reduces Miro1 levels in fibroblasts from Parkinson's disease (PD) patients (IC50 = 7.8 μM). Exhibits no significant effect on related outer mitochondrial membrane protein Mitofusin. Reduces stress-induced degeneration of dopaminergic neurons derived from PD patient iPSCs. Rescues age-dependent neuronal loss and prolongs lifespan in fly PD models.
Specifications
Specifications
| Chemical Name or Material | N-(3-Fluorophenyl)-N'-[4-[[6-(1 H-imidazol-1-yl)-4-pyrimidinyl]amino]phenyl]urea hydrochloride |
| Quantity | 10 mg |
| Target | Rhot1 |
| Molecular Formula | C20H16FN7O.HCl |
| Purity | 0.98 |