Learn More
Sartorius Minisart™ HY PTFE Syringe Filter
BIOPHARMA
Product Code. 17163531 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17163531 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17163531 Supplier Sartorius Supplier No. S6596FMOSK
Only null left
Add to basket
Add to basket
Designed for fast and reliable venting and gas filtration. Minisart™ HY Syringe Filters are single-use, complete filtration units with naturally hydrophobic PTFE membranes and medical acrylic housing.
  • Individually packed and sterile
  • Minisart Standard HY syringe filters Type S6596 with a pore size of 0.2 μm are optimal for immediate usage whenever sterile filtration or venting is needed.
  • Due to its hydrophobic nature, the PTFE membrane functions as a water barrier and does not become wetted by aerosols.
  • This keeps the hydrophobic venting filter permeable for gas or air!
  • The housing of the filter is made of MBS (Methacrylate butadiene styrene) which has a limited compatibility with alcohols but is free of phthalates.
  • Minisart™ Standard Syringe Filters without CE marking are not for medical use.
  • Sterile and ready-to-use ETO sterilized.
  • Suitable as venting filter.
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Filter Type Syringe Filter
For Use With (Equipment) Syringe
Material (Membrane) PTFE (Hydrophobic)
Pore Size 0.2 μm
Sterility Sterile
For Use With (Application) Sterilization and cleaning of air, gases and venting
Hold up Volume (After Air Purge) ≤100 to 150 μm
Housing Material MBS
Inlet Style Female Luer Lock
Outlet Style Male Luer Lock
Packaging Single blister packed
Quantity 50/Pk.
Sterilization Method Ethylene Oxide (ETO)
Diameter (English) 1 in.
Diameter (Metric) 26 mm
Fit for Purpose Sterile