Learn More
Brady™ MiniMark™ Labeling Ribbons
Description
- These ribbons have standard scratch, smear and chemical resistance MiniMark ribbon for B-7951 and B-7589 materials.
- MiniMark ribbon for B-7951 and B-7589 materials.
Specifications
Specifications
| Product Type | Printer Ribbon |
| Color | Blue |
| For Use With (Equipment) | MiniMark Printer |
| Length (English) | 98.4 yd. |
| Width (English) | 4.3 in. |
| Length (Metric) | 90 m |
| Width (Metric) | 110 mm |
| Diameter (English) | 0.5 in. ID |
| Diameter (Metric) | 12.7 mm I.D. |
| Dimensions (L x W) | 90 m x 110 mm |
| Show More |