Learn More
Bryson Packaging™ Polyethylene Bags
missing translation for 'orderingAttributeHoverText'
Width (Metric):
40 mm
60 mm
75 mm
90 mm
100 mm
115 mm
125 mm
150 mm
190 mm
230 mm
255 mm
330 mm
Height (Metric):
65 mm
75 mm
80 mm
115 mm
140 mm
175 mm
190 mm
230 mm
325 mm
355 mm
457 mm
Color:
Clear
Clear with White Bars
missing translation for 'unitSize'
Pack of 100
Description
- Available in a wide range of sizes/capacities
- With resealable strips
Specifications
Specifications
| Description | Resealable Bag |
| Capacity (Metric) | 760 mL |
| Color | Clear |
| For Use With (Application) | Ideal for storing stationery and sending samples |
| Material | Polyethylene |
| Width (Metric) | 115 mm |
| Height (Metric) | 115 mm |