Learn More
Scie Plas Mini - Plusi Gel Electrophoresis Unit
Product Code. 15845541
Change view
Click to view available options
Gel Dimensions:
110.5 x 100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Dimensions unitSize
15845541 110.5 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15845541 Supplier Scie Plas Supplier No. HU10
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for screening PCR products, DNA mini-preps, plasmid vectors, restriction fragments
  • Used for routine preparatory and analytical electrophoresis techniques
  • Four way casting options (either directly within tank or externally when tank is in use)
  • Colour coded, height adjustable combs
  • Four comb positions along tray for swift separation of multiple samples
  • Coloured loading strips for easy well detection when loading
  • Compact tank to reduce buffer volume required to cover gel
  • UV-transparent acrylic casting tray to allow user to image gel
  • Side handles for safe, easy transportation around laboratory
Comb Specifications:
  • Power output connector: Shrouded, 4mm (diameter)
  • Comb thickness: 1, 1.5 or 2mm
  • Comb throughput: 8 to 20 samples
  • No of combs: 2
  • Migration distance between comb slots: 2.5cm
Specifications
Type Gel Electrophoresis Unit
Gel Dimensions 110.5 x 100 mm
Includes 1 x gel casting tray, 2 x 1.0mm 8 sample combs, power supply connectors and loading strips
Model Mini
Sample Size 32
Volume (Metric) Buffer 450 mL
Length (Metric) Gel 11.5 cm
Width (Metric) Gel 10 cm
Voltage 75 to 125 V