Learn More
Cytiva Mini Dialysis Kits
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
482.00€
Specifications
Product Type | Mini Dialysis Kit |
---|---|
Kit Contents | 50 x tube with dialysis cap, 50 x standard cap, 6 x float |
Quantity | Each |
For Use With (Application) | Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane |
Includes: 50 tubes with 50 dialysis and 50 standard caps, plus six floats
Product Code | Brand | Capacity (Metric) | MWCO | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | MWCO | Price | Quantity & Availability | ||||
11520694
|
Cytiva
80-6484-32 |
2 mL | 8 kDa |
482.00€
1 set |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
10196074
|
Cytiva
80-6483-75 |
250 μL | 1 kDa |
482.00€
1 set |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
11510694
|
Cytiva
80-6483-94 |
2 mL | 1 kDa |
482.00€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10617465
|
Cytiva
80-6484-13 |
250 μL | 8 kDa |
482.00€
250µL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
Description
- Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
- Uses disposable dialysis tubes with dialysis membrane incorporated into caps
- Conical tube bottom maximizes sample recovery
- Process complete in as little as several hours
- Choice of molecular weight cut-off and tube size
Specifications
Mini Dialysis Kit | |
50 x tube with dialysis cap, 50 x standard cap, 6 x float | |
Each | |
Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane |