Learn More
Integra™ Miltex™ Biopsy Punches with Plunger System
Product Code. 13404556
Encompass
Change view
Click to view available options
Diameter (Metric):
1 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
13404556 1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13404556 Supplier Integra™ Supplier No. 3331AAP/25
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Effortless ejection of lodged specimens.
Effortless ejection of lodged specimens
  • Seamless, stainless steel with razor-sharp cutting edge to eliminate torn or pulled biopsies
  • Ribbed handle provides comfort and control
  • Single-use
  • Sterile
  • Color coded with size clearly marked on each punch
Order Info
Five-year shelf life
Specifications
Blade Material Stainless Steel
Product Type Biopsy Punch
For Use With (Application) Offer the added benefit of effortless ejection of Lodged Specimens
Handle Type Ribbed
Description Seamless
Disposable Single-use
Sterility Sterile
Includes Plunger System
Quantity 25/Pk.
Diameter (English) 0.039 in.
Diameter (Metric) 1 mm