Learn More
Merck Millipore MILLIPLEX MAP Canine Cytokine/Chemokine Magnetic Bead Panel
MILLIPLEX MAP Canine Cytokine/Chemokine Magnetic Bead Panel enables you to focus on the potential role of cytokines in disease, as well as the modulation of cytokine expression.
Brand: Merck Millipore CCYTMG-90K-PX13
Description
- MILLIPLEX MAP Canine Cytokine/Chemokine Magnetic Bead Panel - Premixed 13 Plex - Immunology Multiplex Assay
- In addition to studying inflammation for canine nutrition and disease research, the dog is an important animal model for human biomedical research and drug development. Millipore recognizes that the availability of canine-specific immunoassay products is limited. Therefore, the MILLIPLE MAP Canine Cytokine / Chemokine Panel enables you to focus on the potential role of cytokines in disease, as well as the modulation of cytokine expression. Coupled with the Luminex™ xMAP™ platform in a magnetic bead format, you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of dozens of analytes simultaneously, dramatically improving productivity.
- The immune system is an organized complex network of biological structures and processes that protect against disease. For example, cytokine and chemokines mediate interactions between cells directly, regulating target immune cell responses. These responses result in inflammation where the immune system attempts to eradicate foreign antigens and begin the healing process. Consequently, the inflammatory process plays a key protective role in immunity. In addition cytokine and chemokine research is essential in understanding the immune system and its multi-faceted response to most antigens, as well as disease states such as autoimmune disease, allergic reactions, sepsis and cancer.
- Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
- Panel Type: MAGNETIC Cytokines/Chemokines
Safety and Handling
- MILLIPLEX Canine Cytokine/Chemokine MAGNETIC Panel Premixed 13 Plex Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Ethylenedinitrilotetraacetic acid,Tetrasodiumsalt, sodium azide