Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
- 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
- Single-well feeder tray utilizes baffles to reduce media leakage and contamination
Specifications
Specifications
| Closure Included | With Lid |
| Bottom Shape | Flat |
| Capacity (Metric) | 22 to 28 mL |
| Color | Clear |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Type | Receiver Tray |
| Well Count | 1 |
| Show More |