Learn More
Merck Millicell μ-Angiogenesis Inhibition Assay Kit
Product Code. 11770363 Shop All Merck Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11770363 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11770363 Supplier Merck Supplier No. MMA125
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
Order Info
dimType
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Fluorescence
For Use With (Application) For Real-Time Monitoring of Changes In Tubule Formation With Unprecedented Optical Resolution
Includes Millicell™ μ-Angiogenesis Slides: Five tissue Culture Treated Slides, Each Slide Contains 15 wells, D, L-Sulforaphane: One 1 mg vial, Calcein AM: One 50 μg vial, One 1 mL vial
Quantity 1 Kit